missing translation for 'onlineSavingsMsg'
Learn More
1,3-Dihydroxynaphthalene, 98%, Thermo Scientific Chemicals Product Code.: 11423338

1,3-Dihydroxynaphthalene, 98%, Thermo Scientific Chemicals

Product Code. 11423338
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11423338

Brand: Thermo Scientific Alfa Aesar A17739.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in the naphthoresorcinol test for uronic acids of Tollens and Rorive, a blue color can be extracted with ethanol after heating of uronic acids with naphthoresorcinol in hydrochloric acid. It is also employed as a reagent for various medical tests. It can also be used as a spot test reagent for glycerate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the naphthoresorcinol test for uronic acids of Tollens and Rorive, a blue color can be extracted with ethanol after heating of uronic acids with naphthoresorcinol in hydrochloric acid. It is also employed as a reagent for various medical tests. It can also be used as a spot test reagent for glycerate.

Solubility
Soluble in ethanol (50 mg/ml), and water.

Notes
Light Sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry and well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-86-5
Molecular Formula C10H8O2
MDL Number MFCD00003965
InChI Key XOOMNEFVDUTJPP-UHFFFAOYSA-N
Synonym 1,3-naphthalenediol, 1,3-dihydroxynaphthalene, naphthoresorcinol, naphthoresoucinol, 3-hydroxybenzocyclohexadien-1-one, naphthoresorcin, ccris 7896, unii-5x457yew8y, acmc-1bvwb, naphthalenediol-1,3Show MoreShow Less
PubChem CID 8601
IUPAC Name naphthalene-1,3-diol

Specifications

Melting Point 123°C to 126°C
Quantity 1 g
Beilstein 2044002
Sensitivity Light sensitive
Merck Index 14,6396
Solubility Information Soluble in ethanol (50mg/ml),and water.
SMILES C1=CC=C2C(=C1)C=C(C=C2O)O
Molecular Weight (g/mol) 160.172
Formula Weight 160.17
Percent Purity 98%
Chemical Name or Material 1,3-Dihydroxynaphthalene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX08K002 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog NumberA17739.06,A17739.03
Lot Number10250598 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog NumberA17739.06,A17739.03
Lot NumberM05K017 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog NumberA17739.06,A17739.03
Lot Number10244068 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberA17739.06,A17739.03
Lot Number10237854 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA17739.06,A17739.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Dihydroxynaphthalene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.