missing translation for 'onlineSavingsMsg'
Learn More
1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride, 90%, Thermo Scientific Chemicals Product Code.: 10085633

1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride, 90%, Thermo Scientific Chemicals

Product Code. 10085633
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10085633

Brand: Thermo Scientific Acros 378340050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 258278-25-0
Molecular Formula C27H39ClN2
Molecular Weight (g/mol) 427.07
MDL Number MFCD07369796
InChI Key LWPXTYZKAWSRIP-UHFFFAOYSA-M
Synonym 1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1h-imidazol-3-ium chloride, sipr.hcl, 1,3-bis 2,6-diisopropylphenyl-imidazolidinium-chloride, 1,3-bis 2,6-diisopropylphenyl imidazolidinium chloride, n,n'-2,6-diisopropylphenyl dihydroimidazolium chloride, 1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1??-imidazol-1-ylium chloride, 1,3-bis 2,6-diisopropylphenyl imidazolinium chloride?, 1,3-bis-2,6-diisopropylphenyl imidazolidinium chloride, 1,3-bis-2,6-diisopropylphenyl imidazolidiniumchloride, 1,3-bis 2,6-bis isopropyl phenyl imidazolin-3-ium chloride
PubChem CID 2734918
IUPAC Name 1,3-bis[2,6-di(propan-2-yl)phenyl]-4,5-dihydroimidazol-1-ium;chloride
SMILES [Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1CC[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 88% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Formula Weight 427.07
Percent Purity 90%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Bis(2, 6-diisopropylphenyl)imidazolidinium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0411202 Certificate TypeCertificate of Analysis Date25/02/2022 Catalog Number378340050,378340010
Lot NumberA0372306 Certificate TypeCertificate of Analysis Date06/05/2021 Catalog Number378340050,378340010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride, 90%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.