Stability Testing of New Drug Substances and Products Q1A (R2) with Memmert
ICH guideline Q1A (R2) for registration purposes and emphasising topics such as stability testing, forced degradation studies and storage testing in combination with products to be used.
Watch the ReplayAgenda
This is a guideline developed by ICH Expert Working Group. It promotes health through international harmonisation and defines the stability data package. This package for a new drug is capable for registration application in EC, Japan and the U.S.A.
A stability test shows drug product/substance testing with environmental factor variables - temperature, humidity and light.
It determines the pharmaceutical product's response to any physical, chemical or microbiological changes. It helps establish the integrity and efficiency of the final product and is important for establishing a re-test period for the drug substance or a shelf life for the drug product and recommended storage conditions.
Details on stress testing and storage condition - long term, intermediate, accelerated - are covered and most suitable products for stability testing are introduced.
Webinar key topics:
- ICH Guideline Q1A (R2)
- Stability testing - forced degradation studies - storage testing
- Climate chambers
Related documents
- Whitepaper: Stability Tests According to ICH Q1A (R2)
- Flyer: Constant Climate Chamber HPP Eco
- Brochure: Climate chambers
Our Speaker
Andrea Weiss
Senior Area Manager, Memmert.
Register now to watch the webinar
Related Webinars