accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now
Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 58438-03-2

CAS RN 58438-03-2

IUPAC Name: (2S)-2-amino-3-(naphthalen-2-yl)propanoic acid
Synonyms: β-(2-naphthyl)-alanine
Molecular Weight (g/mol): 215.25
Molecular Formula: C13H13NO2
InChi Key: JPZXHKDZASGCLU-LBPRGKRZSA-N
SMILES: N[C@@H](CC1=CC=C2C=CC=CC2=C1)C(O)=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.