man

Test

CAS RN 5608-24-2

OHHONO

CAS RN 5608-24-2

IUPAC Name: 2,2-bis(hydroxymethyl)-1-azabicyclo[2.2.2]octan-3-one
Synonyms: 2,2-bis(hydroxymethyl)-1-azabicyclo[2.2.2]octan-3-one
Molecular Weight (g/mol): 185.22
Molecular Formula: C9H15NO3
InChi Key: RFBVBRVVOPAAFS-UHFFFAOYSA-N
SMILES: OCC1(CO)N2CCC(CC2)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results