accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 105942-08-3

CAS RN 105942-08-3

IUPAC Name: 4-bromo-2-fluorobenzonitrile
Synonyms: 4-bromo-2-fluorobenzonitrile
Molecular Weight (g/mol): 200.01
Molecular Formula: C7H3BrFN
InChi Key: HGXWRDPQFZKOLZ-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC=C1C#N
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.