accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 105816-04-4

CAS RN 105816-04-4

IUPAC Name: (2R)-3-phenyl-2-{[(1S,3S)-3-(propan-2-yl)cyclohexyl]formamido}propanoic acid
Synonyms: nateglinide
Molecular Weight (g/mol): 317.43
Molecular Formula: C19H27NO3
InChi Key: CZCVBLROPSHDRZ-YESZJQIVSA-N
SMILES: CC(C)[C@H]1CCC[C@@H](C1)C(=O)N[C@H](CC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.