accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 105728-90-3

CAS RN 105728-90-3

IUPAC Name: 2-fluoro-5-methoxybenzaldehyde
Synonyms: 2-fluoro-5-methoxybenzaldehyde
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: DKIQXHIAEMGZGO-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C(C=O)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.