accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10537-47-0

CAS RN 10537-47-0

IUPAC Name: 2-[(3,5-di-tert-butyl-4-hydroxyphenyl)methylidene]propanedinitrile
Synonyms: 2-[(3,5-di-tert-butyl-4-hydroxyphenyl)methylidene]propanedinitrile
Molecular Weight (g/mol): 282.39
Molecular Formula: C18H22N2O
InChi Key: MZOPWQKISXCCTP-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(C=C(C#N)C#N)=CC(=C1O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.