accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10526-80-4

CAS RN 10526-80-4

IUPAC Name: 2-(phosphonooxy)prop-2-enoic acid; cyclohexanamine
Synonyms: cyclohexylamine; phosphoenolpyruvic acid
Molecular Weight (g/mol): 267.22
Molecular Formula: C9H18NO6P
InChi Key: VHFCNZDHPABZJO-UHFFFAOYSA-N
SMILES: NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.