accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10504-60-6

CAS RN 10504-60-6

IUPAC Name: methyldiphenylsulfanium; tetrafluoroboranuide
Synonyms: methyldiphenylsulfanium tetrafluoroborate
Molecular Weight (g/mol): 288.11
Molecular Formula: C13H13BF4S
InChi Key: LHAMVDBAOJCBDW-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.C[S+](C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.