accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 104934-52-3

CAS RN 104934-52-3

IUPAC Name: 3-dodecylthiophene
Synonyms: 3-dodecylthiophene
Molecular Weight (g/mol): 252.46
Molecular Formula: C16H28S
InChi Key: RFKWIEFTBMACPZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CSC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.