accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1047626-77-6

CAS RN 1047626-77-6

IUPAC Name: 2-iodo-1-(oxan-2-yl)-1H-imidazole
Synonyms: 2-iodo-1-(oxan-2-yl)imidazole
Molecular Weight (g/mol): 278.09
Molecular Formula: C8H11IN2O
InChi Key: UWFGOSIGMHYERY-UHFFFAOYNA-N
SMILES: IC1=NC=CN1C1CCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.