CAS RN 103457-72-3
CAS RN 103457-72-3
IUPAC Name:
erbium(3+) tris(bis(trimethylsilyl)azanide)
Synonyms:
erbium(3+) tris(bis(trimethylsilyl)azanide)
Molecular Weight (g/mol):
648.42
Molecular Formula:
C18H54ErN3Si6
InChi Key:
ZVNGWTZZRANJAO-UHFFFAOYSA-N
SMILES:
[Er+3].C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C
1
–
2
of
2
results
2
Tris[N,N-bis(trimethylsilyl)amide]erbium(III), 98%, Thermo Scientific Chemicals
CAS: 103457-72-3 Molecular Formula: C18H54ErN3Si6 Molecular Weight (g/mol): 648.42 MDL Number: MFCD00271002 InChI Key: ZVNGWTZZRANJAO-UHFFFAOYSA-N Synonym: bis trimethylsilyl azanide; erbium 3+,tris n,n-bis trimethylsilyl amide erbium iii,acmc-20al2x,erbium iii bis trimethylsilyl amide,tris bis trimethylsilyl amino erbium,erbium tris bis trimethylsilyl amide,erbium 3+ tris bis trimethylsilyl azanide,erbium tris trimethyl-n-trimethylsilyl silanaminide,silanamine,1,1,1-trimethyl-n-trimethylsilyl-, erbium 3+ salt 3:1 PubChem CID: 6098739 IUPAC Name: bis(trimethylsilyl)azanide;erbium(3+) SMILES: C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Er+3]