missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris[N,N-bis(trimethylsilyl)amide]erbium(III), 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Specifications
Specifications
CAS | 103457-72-3 |
Melting Point | 180°C to 184°C |
Color | Yellow to Orange |
Elemental Analysis | 32.5 to 34 % (C) 8.2 to 8.6 % (H) |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | C18H54ErN3Si6 |
Linear Formula | Er[N(Si(CH3)3)2]3 |
MDL Number | MFCD00271002 |
Quantity | 1 g |
Show More |
Safety and Handling
Product Identifier
- Tris¢N,N-bis(trimethylsilyl)amide!erbium(III)
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 2
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H261-In contact with water releases flammable gases.
- H228-Flammable solid.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P223-Do not allow contact with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402-Store in a dry place.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'