accessibility menu, dialog, popup

Test

CAS RN 10026-13-8

CAS RN 10026-13-8

IUPAC Name: pentachloro-λ⁵-phosphane
Synonyms: phosphorus pentachloride PCl5
Molecular Weight (g/mol): 208.22
Molecular Formula: Cl5P
InChi Key: UHZYTMXLRWXGPK-UHFFFAOYSA-N
SMILES: ClP(Cl)(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)