missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus pentachloride, 98%, Thermo Scientific Chemicals

Product Code. 10793144
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-13-8
Molecular Formula Cl5P
MDL Number MFCD00011439
InChI Key UHZYTMXLRWXGPK-UHFFFAOYSA-N
Synonym unii-0ex753tydu, pcl5, phosphorane, pentachloro, phosphorus perchloride, phosphorus pentachloride, phosphorous pentachloride, phosphoric perchloride, phosphorpentachlorid, phosphorus v chloride, pentachlorophosphorane
PubChem CID 24819
ChEBI CHEBI:30335

Specifications

Color Yellow
Physical Form Crystalline Powder and Scales
Quantity 2.5 kg
Assay Percent Range 98%
Linear Formula PCl5
Fieser 01,866; 04,388; 05,534; 07,290
SMILES ClP(Cl)(Cl)(Cl)Cl
IUPAC Name pentachloro-λ⁵-phosphane
Molecular Weight (g/mol) 208.22
Formula Weight 208.24
Percent Purity ≥97.5%
Packaging Glass bottle
Density 1.6 g/mL
Chemical Name or Material Phosphorus pentachloride, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phosphorus pentachloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Phosphorus pentachloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.