accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10025-82-8

ClClClIn

CAS RN 10025-82-8

IUPAC Name: indium(3+) trichloride
Synonyms: indium(III) chloride
Molecular Weight (g/mol): 221.17
Molecular Formula: Cl3In
InChi Key: PSCMQHVBLHHWTO-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[In+3]
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
Physical Form 
  • (5)
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (4)
Grade 
  • (3)

Narrow Results

Narrow Results

  • (3)
  • (12)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (4)
  • (3)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.