accessibility menu, dialog, popup

Test

CAS RN 10025-77-1

ClClClFeH2OH2OH2OH2OH2OH2O

CAS RN 10025-77-1

IUPAC Name: hexahydroxyirontris(ylium) trichloride
Synonyms: hexaaquairon(3+) trichloride
Molecular Weight (g/mol): 270.29
Molecular Formula: Cl3FeH12O6
InChi Key: NQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
Quantity 
  • (8)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
Percent Purity 
  • (4)
  • (4)
  • (11)
  • (7)
  • (3)
Grade 
  • (4)
  • (8)
  • (3)
  • (4)
  • (4)
Physical Form 
  • (3)
  • (1)
  • (4)
  • (1)
  • (8)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (11)
  • (4)
  • (2)
  • (1)
  • (2)
  • (11)
  • (7)
  • (1)
  • (1)
  • (8)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (4)
  • (4)
  • (11)
  • (7)
  • (3)
  • (4)
  • (8)
  • (3)
  • (4)
  • (4)
  • (3)
  • (1)
  • (4)
  • (1)
  • (8)
  • (6)
  • (2)