missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) Chloride, 4.5% Solution, Chem-Lab
Click to view available options
Quantity:
100 mL
Description
Iron(III) chloride hexahydrate is employed as a Lewis acid for catalyzing reactions and for colorimetric testing for phenols and their derivatives. It is used as a clarifying agent in wastewater and drinking water treatment. Other uses of Iron(III) chloride hexahydrate include chloride hydrometallurgy and for the detection of gamma-hydroxybutyric acids and and salicylic acids.
Specifications
Specifications
Melting Point | 0°C |
CAS | 10025-77-1 |
Color | Yellow |
pH | <1 |
Physical Form | Liquid |
Quantity | 100 mL |
Molecular Formula | FeCl3 • 6H2O |
MDL Number | MFCD00149712 |
Synonym | Ferric chloride hexahydrate, Trichloroiron hexahydrate, Iron trichloride |
Solubility Information | Soluble in Water |
Show More |
Safety and Handling


Product Identifier
- Iron(III) chloride 4.5% solution (Yellow sol.)
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P313-Get medical advice/attention.
- MIXTURE LIST-Contains : iron(III) chloride,hydrochloric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction