accessibility menu, dialog, popup

Test

CAS RN 100-09-4

H3COOHO

CAS RN 100-09-4

IUPAC Name: 4-methoxybenzoic acid
Synonyms: P-anisic acid P-methoxybenzoic acid p-isomer anisic acid draconic acid 14C-carboxy 4-anisic acid 4-anisic acid, 14C-carboxy copper (+2) salt 4-anisic acid anisic acid, p- anisic acid, p-isomer kyselina 4-methoxybenzoova
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: ZEYHEAKUIGZSGI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(O)=O
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)