missing translation for 'onlineSavingsMsg'
Learn More

p-Anisic acid, 98%, Thermo Scientific Chemicals

Product Code. 13418378
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-09-4
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002542
InChI Key ZEYHEAKUIGZSGI-UHFFFAOYSA-N
Synonym p-anisic acid, anisic acid, p-methoxybenzoic acid, draconic acid, 4-anisic acid, benzoic acid, 4-methoxy, anisic acid, para, 4-methoxybenzoate, para-anisic acid, anisic acid, p-isomer
PubChem CID 7478
ChEBI CHEBI:40813
IUPAC Name 4-methoxybenzoic acid
SMILES COC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 181.0°C to 186.0°C
Color Beige-Gray to White
Boiling Point 275.0°C
Flash Point 185°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic drum
Linear Formula CH3OC6H4CO2H
Refractive Index 1.5710 to 1.5760
Quantity 5 kg
Beilstein 10,IV,346
Merck Index 15,659
Solubility Information Solubility in water: 0.3g/L (20 c). Other solubilities: freely soluble in alcohol,ether,chloroform and,ethyl acetate
Formula Weight 152.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material p-Anisic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.