Learn More
RETSCH Zirconium Oxide Comfort Grinding Jars
Use these grinding jars with Retsch™ ball mills. Retsch™ Zirconium Oxide Comfort Grinding Jars feature an O-type sealing for safe operation.
Brand:  RETSCH 01.462.0219
 View more versions of this product
Product Code. 10671083
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Available in a variety of sizes
  • Optional aeration covers
  • Pressure tight safety closure devices
Specifications
Specifications
Comfort Grinding Jar
250 mL
For PM 100, PM 400 and S 100
Zirconium Oxide
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products