missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Zinc Test Kits
Get optimum zinc measurement results in waste water, particularly from electroplating and metal processing industries, seepage water, sewage sludge, soils with CombiCheck Zinc (Zn) cell test kits.
Brand: WTW™ 250417
This item is not returnable.
View return policy
Description
- CombiCheck Zinc (Zn) cell test kit
- For use with PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
- For waste water, particularly from electroplating and metal processing industries, seepage water, sewage sludge, soils
Specifications
PAR | |
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers | |
25 | |
Photometric |
For waste water, particularly from electroplating and metal processing industries, seepage water, sewage sludge, soils | |
Certificates | |
25 Tests | |
CombiCheck Zinc (Zn) cell test kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Zinc Cell Test Method: photometric 0.20 - 5.00 mg/l Zn Spectroquant
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Potassium cyanide