missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc sulfate heptahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
6 x 500 g
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
CAS Max % | 100.0000% |
Quantity | 6 x 1 kg |
Linear Formula | ZnSO4 · 7H2O |
Formula Weight | 287.56 |
Percent Purity | >99.5% |
Grade | ACS Reagent/Reagent ISO/Reagent Ph. Eur. |
Packaging | Poly Bottle |
Density | 1.96 g/cm3 |
Chemical Name or Material | Zinc sulfate heptahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'