missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.0-103.0%, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
CAS Max % | 100.0000% |
Quantity | 25 kg |
Assay Percent Range | 99 to 103% |
Linear Formula | ZnSO4 · 7H2O |
UN Number | UN3077 |
Formula Weight | 287.56 |
Percent Purity | 99.0 to 103.0% |
Grade | puriss. |
Packaging | Paper / Poly Bag |
Density | 1.96 g/cm3 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'