missing translation for 'onlineSavingsMsg'
Learn More

Zinc powder, average 5-8 micron, 97.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11356967
Click to view available options
Quantity:
100 g
1 kg
5 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Appearance Gray powder
Density 7.14
Assay Percent Range 97.5% (Metals basis)
Quantity 100 g
Odor Odorless
Sensitivity Air and moisture sensitive
Form Powder/Flakes
Chemical Name or Material Zinc

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'