missing translation for 'onlineSavingsMsg'
Learn More

Zinc Oxide, Honeywell Fluka™

Product Code. 15611430
Click to view available options
Quantity:
100 g
1 kg
6 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5% (calc. for dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Quantity 100 g
Linear Formula ZnO
UN Number 3077
Formula Weight 81.39g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Chemical Name or Material Zinc Oxide, Meets analytical specification of Ph. Eur.BP, USP
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'