missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ZD 7155 hydrochloride
Selective non-peptide AT1 antagonist
233.00€ - 906.00€
Specifications
Chemical Name or Material | 5,7-Diethyl-3,4-dihydro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1,6-naphthyridin-2(1H)-one hydrochloride |
---|---|
CAS | 146709-78-6 |
Assay Percent Range | >98% |
Solubility | Soluble to 25mM in ethanol |
Molecular Formula | C26H26N6O.HCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18015922
|
Tocris Bioscience™
1211/10 |
10 mg |
233.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18086032
|
Tocris Bioscience™
1211/50 |
50 mg |
906.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A potent and selective competitive antagonist for the angiotensin II type 1 (AT1) receptor. Displaces [125I]-angiotensin II binding with an IC50 value of 3.8 nM in guinea pig adrenal gland membranes. Orally active, and is more potent and longer-acting than the prototype AT1 antagonist, losartan.Specifications
5,7-Diethyl-3,4-dihydro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1,6-naphthyridin-2(1H)-one hydrochloride | |
>98% | |
C26H26N6O.HCl |
146709-78-6 | |
Soluble to 25mM in ethanol | |
Observed MW: 474.99 |
Safety and Handling
Recommended Storage : Desiccate at +4°C
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title