missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB7A/Pokemon Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-761AF594
This item is not returnable.
View return policy
Description
ZBTB7A/Pokemon Polyclonal antibody specifically detects ZBTB7A/Pokemon in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| ZBTB7A/Pokemon | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| DKFZp547O146, Factor binding IST protein 1, Factor that binds to inducer of short transcripts protein 1, FBI-1POK erythroid myeloid ontogenic factor, FBI1TTF-I-interacting peptide 21, HIV-1 inducer of short transcripts binding protein, Leukemia/lymphoma-related factor, LRFHIV-1 1st-binding protein 1, lymphoma related factor, MGC99631, pokemon, TIP21, ZBTB7zinc finger and BTB domain containing 7, zinc finger and BTB domain containing 7A, zinc finger and BTB domain containing 7A, HIV-1 inducer of short transcriptsbinding protein, zinc finger and BTB domain-containing protein 7A, Zinc finger protein 857A, ZNF857APOZ and Krueppel erythroid myeloid ontogenic factor | |
| The immunogen recognized by this antibody maps to a region between residues 200 and 250 of human ZBTB7 (Factor Binding IST protein 1) using the numbering given in entry NP_056982.1 (GeneID 51341). | |
| 0.1 mL | |
| Cancer, Transcription Factors and Regulators | |
| 51341 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction