Learn More
Thermo Scientific Chemicals Yttria Stabilized Zirconium Oxide Cylindrical Crucibles
Product Code. 15453825
Click to view available options
Capacity (Metric):
5 mL
10 mL
50 mL
100 mL
200 mL
Diameter (Metric) Outer:
20mm
24mm
37mm
45mm
55mm
Height (Metric):
37mm
43mm
72mm
90mm
112mm
Unit Size:
Each
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15453825
Brand: Thermo Scientific Chemicals 041273.KF
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Material Zirconium Oxide
Quantity 1 Piece
Capacity (Metric) 5 mL
Diameter (Metric) Outer 20mm
Height (Metric) 37mm
Product Type Yttria Stabilized Zirconium Oxide Cylindrical Crucibles