Learn More
YSI™ Probe Service Kits for YSI Dissolved Oxygen Probes
For replenishing oxygen sensor on YSI Dissolved Oxygen Probes
Brand: YSI™ 098094
This item is not returnable.
View return policy
Description
- Membrane Kits include 150 standard, 1mil PTFE membranes or 150 high-sensitivity, 0.5mil PTFE membranes, in 10 packs of 50
- Membrane/KCI Kits include 30 standard or high-sensitivity membranes, 1 oz. (30mL) dropper bottle with KCI crystals for adding distilled water and a probe contour tool
- For use with probes 13-299-43 (Model 5739), 13-298-80 (Model 5750), 15-176-50 (Model 5718), 13-298-16 (Model 5300 Biological Oxygen Monitor), and 13-298-56, -57, or -62 (Model 55 Handheld Dissolved Oxygen Meters)
Specifications
5718, 5739, 5750, 5300 and Rapid Pulse Probes | |
Bottle of electrolyte, 2 O-rings, 30 1mL FEP Fluoropolymer Film Membranes | |
Each |
Standard Membrane Kit: 30 membranes, KCl | |
DO |