Learn More
YSI™ Pro Plus Quatro Field Cable
Product Code. 11968089
Click to view available options
:
Pro Plus Quatro Cable; 30m
Product Code. 11968089
Brand: YSI™ 60579030
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Includes
Conductivity/temperature sensor
For use with Professional Plus instrument.
  • 4-port cable
  • True field cable with MS (military spec) connectors
  • Rugged, weighted probe guard
  • Measure conductivity/temperature, dissolved oxygen and any two ISEs among pH, ORP, ammonium, nitrate or chloride
  • Cable Length: 30m
Warranty
Two years
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Professional Plus instrument