accessibility menu, dialog, popup
Test
chevron-down
Select Language
English
French
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
pH and Electrochemistry
Probes and Electrodes
Conductivity Electrodes
YSI™ Flow Cell Mounting Spike
Print
YSI™ Flow Cell Mounting Spike
410.00€
Specifications
Type
Flow Cell Mounting Spike
View More Specs
Products
1
Documents
Product Certifications
Products
1
Product Code
Brand
For Use With (Equipment)
Includes
Price
Quantity & Availability
Product Code
Brand
For Use With (Equipment)
Includes
Price
Quantity & Availability
11978736
View Documents
missing translation for 'viewProductCertificates'
YSI™
603076
3059, 3077 flow cells
Extended length
410.00€
Each
In Stock
Log in
to see stock.
Specifications
Type
Flow Cell Mounting Spike
Documents
Product Certifications
Back to Top