Learn More
Trajan Scientific and Medical™ XCHANGE™ CTC PAL-xt Autosampler Syringes
Product Code. p-7114379
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
Diameter (Metric) Outer Needle:
0.63 mm
0.71 mm
Material:
Metal (Plunger)
PTFE (Plunger)
PTFE-tipped (Plunger)
Unit Size:
Each
Product Code. 15249263
Brand: Trajan Scientific and Medical™ 2928310
Allows adding of internal standards without cross contamination
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Available as all-metal or PTFE-tipped plunger
Specifications
Material PTFE (Plunger)
Description Autosampler Syringe
Needle Gauge 23 G
Diameter (Metric) Inner Needle 0.15 mm
Diameter (Metric) Outer Needle 0.63 mm
Volume (Metric) 25 μL
Needle Point Style Cone