Learn More
Thermo Scientific Matrix White color inserts for 500 µl and 1.0ml
Brand:  Thermo Scientific Matrix 4475-11
22.41 EUR valid until 2024-09-27
Use promo code "21650" to get your promotional price.
White color inserts for 500 µl and 1.0ml
Product Code. 10000273
  • 30.70€ / Pack of 500
Estimated Shipment: 14-11-2024