Learn More
ABEBA™ X-Light White Occupational Shoes O2 ESD
White X-light occupational shoes, with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, PU slip resistant outsole
Brand:  ABEBA™ 7131124/43
 View more versions of this product
Product Code. 17141505
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Lightweight, comfortable white occupational shoes
  • Comply to EN ISO 20347: 2012, O2 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant Mono PU outsole SRC
  • White technical fabric
  • Silver ions lining
  • Replaceable active comfort insole
Specifications
Specifications
White
43
Unisex
Water Resistant
Synthetic fabric
Comply to EN 20347: 2012 O2 SRC, ESD dissipative according EN 61340
Low-cut
O2 SRC ESD