Learn More
Tuttnauer Wire (Mesh) Basket
Shop All Tuttnauer Products
Click to view available options
Capacity (Metric):
19.5 L
26 L
29 L
Dimensions (Dia. x H):
362 x 190 mm
362 x 250 mm
362 x 280 mm
Unit Size:
Each
Description
- Stainless Steel wire basket for the sterilization of solid items, or liquid in bottles.
- The baskets are stackable and two baskets can be placed in the autoclave chamber.
Specifications
Specifications
| Capacity (English) | 6.4 gal. |
| Capacity (Metric) | 29 L |
| Depth (Metric) Chamber | 28 cm |
| Depth (English) Chamber | 11.2 in. |
| Length (English) | 14.3 in. |
| Length (Metric) | 36.2 cm |
| Product Type | Autoclave Basket |
| Dimensions (Dia. x H) | 362 x 280 mm |
| Sterility | Non-sterile |