Learn More
Bel-Art™ Wilmad™ Rotor and Caps for Bruker MAS Probe
Product Code. p-6960071 Shop All SP Scienceware Products
Click to view available options
Model:
WP-501-2180-SET1
WP-501-3180
WP-501-3180-SET1
WP-501-4180-SET-5
WP-501-7180-SET-5
WP-601-4181
WP-602-3181
Type:
Cap
Probe
Rotor
For Use With (Equipment):
Bruker MAS Probe (2.5 mm)
Bruker MAS Probe (3.2 mm)
Bruker MAS Probe (4.0 mm)
Bruker MAS Probe (7.0 mm)
Unit Size:
Each
Product Code. 15388678
Brand: Bel-Art™ 660000096
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Eliminate the need to solvate with MAS rotors manufactured from the highest purity Zirconia. Rotors provide the ultimate solution for analysis of solid samples.
  • Caps fitted with O-rings for improved sealing
  • MAS Probe and caps are 100% compatible for most solid state NMR spectrometers
  • Thoroughly inspected before and after the precision machining process to ensure there are no material irregularities
  • Spin testing is performed to only the highest specified spinning speed, assuring performance without overspinning the rotor
Specifications
For Use With (Equipment) Bruker MAS Probe (2.5 mm)
Type Probe
Description One 2.5mm Rotor, Two Vespel Caps and Botoms
Material Various
Model WP-501-2180-SET1
Speed Vmax = 35 kHz
Temperature Range (Metric) -150°C to +200°C