Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Varian Spinner Turbine
Product Code. 15368628 Shop All SP Scienceware Products
Click to view available options
For Use With (Equipment):
Agilent/Varian NMR Spectrometer
Model:
V-PEEK-3-NS
Unit Size:
Each
Product Code. 15368628
Brand: Bel-Art™ SP Scienceware™ 660000058
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Varian Spinner Turbine is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
  • Cost effective compared to the full VT version for sample changers
  • Allows a portion of the VT air to pass straight past the sample inside the spinner turbine therefore reducing VT gradients and micro sonic flutter due to a high VT flow
  • Includes high temperature external O-rings at the top and bottom for long life and a firm grip on the sample at top and bottom.
  • Can also be used for spinning experiments.
Specifications
For Use With (Equipment) Agilent/Varian NMR Spectrometer
Includes PEEK exchangeable fingers
Model V-PEEK-3-NS