accessibility menu, dialog, popup
Test
chevron-down
Select Language
English
French
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Specialty Lab Equipment, Instruments, and Apparatuses
Mills
RETSCH Wide Mouth Bottles
10598171
Print
RETSCH Wide Mouth Bottles
Capacity: 500mL
Brand:
RETSCH 22.523.0002
View more versions of this product
Product Code.
10598171
98.35€
/ Pack of 10
Quantity
Estimated Shipment: 27-12-2024
Log in
to see stock.
Explore more special offers
Specifications
Product Type
Wide Mouth Bottle
Capacity (Metric)
500 mL