accessibility menu, dialog, popup
Test
chevron-down
Select Language
English
French
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Specialty Lab Equipment, Instruments, and Apparatuses
Mills
RETSCH Wide Mouth Bottles
10222501
Print
RETSCH Wide Mouth Bottles
Capacity: 250mL
Brand:
RETSCH 22.523.0001
View more versions of this product
Product Code.
10222501
99.50€
/ Pack of 10
Quantity
Estimated Shipment: 13-12-2024
Log in
to see stock.
Explore more special offers
Specifications
Product Type
Wide Mouth Bottle
Capacity (Metric)
250 mL