Learn More
Cytiva Whatman™ Three-piece Filter Funnel
Features quick and easy filtration. Whatman™ Three-piece Filter Funnels dismantle quickly for the insertion of a new filter. Funnels are available with a choice of three plates and come in several sizes.
247.00€ - 720.00€
Product Code | Brand | Diameter (Metric) Membrane | Volume (Metric) | Filtration Area | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Membrane | Volume (Metric) | Filtration Area | Price | Quantity & Availability | |||
11300634
|
Cytiva
1950-002 |
25 mm | 16 mL | 2 cm2 |
247.00€
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||
11310634
|
Cytiva
1950-004 |
47 mm | 36 mL | 8 cm2 |
280.00€
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||
10387671
|
Cytiva
1950-007 |
70 mm | 115 mL | 19.6 cm2 |
339.00€
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||
11330634
|
Cytiva
1950-009 |
90 mm | 200 mL | 38.5 cm2 |
390.00€
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||
11340634
|
Cytiva
1950-012 |
125 mm | 530 mL | 66.5 cm2 |
720.00€
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||
10384061
|
Cytiva
1950-017 |
70 mm | 210 mL | 19.6 cm2 |
389.00€
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||
10184161
|
Cytiva
1950-027 |
70 mm | 400 mL | 19.6 cm2 |
590.00€
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
Description
Features quick and easy filtration. Whatman™ Three-piece Filter Funnels dismantle quickly for the insertion of a new filter. Funnels are available with a choice of three plates and come in several sizes.
The increased use of high-efficiency glass microfiber filters in modern laboratories has created a demand for simple and effective filter-holding systems. Whatman 3-piece filter funnels have been designed to complement the range of Whatman fine particle retention, rapid flow rate glass microfiber filters.- Three-piece construction; funnel is quickly dismantled for insertion of a new filter
- Glass sealing flanges are ground flat to ensure a good filter seal
- Retained solids are deposited within the filter circle
- Edge clamping prevents peripheral loss and possible passage of solution around, rather than through, the filter circle
- Parts can be quickly and efficiently cleaned
- Acrylic - Supplied as standard; suitable for filtration of most aqueous solutions; maximum working temperature 65°C
- Polypropylene Plate - Optional extra. Recommended for most acids (except concentrated nitric acid and fuming sulfuric acid) at room temperatures; suitable also for most alcohols, glycols, ethers and ketones; maximum working temperature 100°C
- PTFE - For virtually all common acids, alkalis and solvents at temperatures up to 100°C; maximum working temperature 200°C
Specifications
Polypropylene | |
Acrylic plate | |
Filter Funnel |