Learn More
Cytiva Whatman™ GV 100 Series Glass Vacuum Filtration Manifolds, Replacement Parts
Click to view available options
Model:
GV 100/0/03
GV 100/0/05
GV 100/1/05
Type:
Centering Ring
Funnel (500 mL)
Glass Frit Filter
Lower Base Support
Material:
Borosilicate Glass
PTFE
For Use With (Equipment):
Manifold Model GV 100/0
Manifold Model GV 100/0 and GV 100/1
Manifold Model GV 100/1
Unit Size:
Each
Description
- Description: Replacement Part for GV 100 Series Glass Vacuum Filtration Manifolds
- For Use With (Application): Vacuum Filtration
Specifications
Specifications
| For Use With (Equipment) | Manifold Model GV 100/0 |
| Material | PTFE |
| For Use With (Application) | Replacement Part for GV 100 Series Glass Vacuum Filtration Manifolds |
| Model | GV 100/0/05 |
| Quantity | 1 Ea. |
| Series | GV100 |
| Type | Centering Ring |