missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Welding Cut Gloves

Product Code. 12611113
Click to view available options
Size:
7
8
9
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Heatproof and water-repellent cowhide are combined with cut and thermally resistant Kevlar™ for durability, resistance, and comfort. Honeywell Safety Products™ Welding Cut Gloves are designed with a protective cuff and an elasticized wrist for a good, protective fit.

  • Suitable for MIG and ARC welding in dry or light oily conditions with cut hazards.
  • Water repellent, heatproof cowhide grain leather back.
  • Water-repellent, heatproof velvet cowhide palm.
  • Water repellent, heatproof split leather cuff to protect the forearm.
  • U.S. assembly.
  • Gunn cut, winged thumb with cord, Crispin type.
  • Elasticated at the back of the hand.
  • Kevlar™ cotton lining for comfort and thermal insulation. Outstanding comfort and flexibility.
TRUSTED_SUSTAINABILITY

Specifications

Back Material Cowgrain leather
Certifications/Compliance CE CATEGORY II Intermediary risks, NF EN 420-03 EEC Basic standard, NF EN 388-03 Mechanical risks, NF EN 407-04 Thermical risks, EN 12477-02 Type A Welding risks, EC Category PPE 2, EC Certificate Number 0075/797/162/10/12/0763, EC Attestation Number 0075/797/162/10/12/0763
Classification Dexterity: 5
Cuff Material Split leather
Cut Style Gunn
Cut Resistance Rating 2
Material Cowhide Leather
Product Type Crispin Welding Gloves (Type A)
Size 7
Thickness (Metric) 0.8/1.1 mm (Back), 1.1/1.3 mm (Cuff)
For Use With (Application) Automotive, Building, Construction, Iron, Steel, Metal, Ship Building and Welding industries
Glove Cuff Style Gauntlet
Liner Material Dupont™ Kevlar™ Aramid Fiber Cotton
Lining Dupont™ Kevlar™ aramid fiber, Cotton
Puncture Rating 2
Quantity 10Pair/PK
Resistant To Heat
Tear Rating 2
Thumb Style Winged with Yoke
Length (Metric) 39 cm
Length (Metric) Cuff 15 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'