missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WedgeWell™ Empty Gel Cassettes with Combs, midi

Product Code. p-200062275
Click to view available options
No. of Wells:
12 + 2-well
20-well
26-well
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30131731

Brand: Invitrogen™ WW1012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These WedgeWell empty gel cassettes are pre-assembled, single-use, plastic cassettes with combs that allow you to pour your own midi-size gels. The gels can be run in the SureLock Tandem Midi Gel Tank (STM1001) or XCell4 SureLock Midi-Cell (WR0100).

These WedgeWell empty gel cassettes are pre-assembled, single-use, plastic cassettes with combs that allow you to pour your own midi-size gels. The gels can be run in the SureLock Tandem Midi Gel Tank (STM1001) or XCell4 SureLock Midi-Cell (WR0100). The cassettes are sealed on three sides to prevent leaking and do not require the use of a special casting stand. Gels poured in these cassettes are 1.0-mm thick. SureCast reagents can used to make pouring your own high performance polyacrylamide midi gels both easy and reproducible.

These WedgeWell empty gel cassettes are pre-assembled, single-use, plastic cassettes with combs that allow you to pour your own midi-size gels. The gels can be run in the SureLock Tandem Midi Gel Tank (STM1001) or XCell4 SureLock Midi-Cell (WR0100). The cassettes are sealed on three sides to prevent leaking and do not require the use of a special casting stand. Gels poured in these cassettes are 1.0-mm thick. SureCast reagents can used to make pouring your own high performance polyacrylamide midi gels both easy and reproducible.

Product features

  • The cassettes are sealed on three sides to prevent leaking and do not require the use of a special casting stand
  • Gels poured in these cassettes are 1.0-mm thick
  • SureCast reagents can be used to pour high-performance polyacrylamide midi gels

Handcast gel reagents

SureCast protein gels are a reliable and high-performance solution for your handcast gel electrophoresis needs. SureCast resolving and stacking buffers are based on the traditional Laemmli protein chemistry, allowing the use of Laemmli sample and running buffers. Handcast SureCast gels are a reliable, high-performance, low-cost option for gel electrophoresis.

Convenient to use

The midi WedgeWell empty gel cassettes have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, which makes sample loading easier.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10 cassettes (with 12+2 well combs) per pack
For Use With (Equipment) XCell4 SureLock Midi-Cell
Gel Size Midi
Type Empty Gel Cassette
No. of Wells 12 + 2-well
Quantity 1 pack
Running Dimension Vertical
Shipping Condition Room Temperature
Thickness (Metric) 1.0 mm
Length (Metric) Gel 8 cm
Width (Metric) 13 cm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.