missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Waymouth's Medium
Waymouth's Medium
43.20€ - 381.00€
Specifications
Certifications/Compliance | ISO 13485 standards, FDA |
---|---|
Content And Storage | 2 to 8°C |
Description | Waymouth's Medium, 500mL, Liquid Form, With L-glutamine, Phenol Red, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture |
Form | Liquid |
Product Type | Waymouth's Medium |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11574426
|
Gibco™
31220023 |
500 mL |
43.20€
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11584426
|
Gibco™
31220072 |
10 x 500 mL |
381.00€
5000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Waymouth's Medium is a chemically defined medium developed by Charity Waymouth and originally formulated for studies on the nutrition, metabolism, and growth of mouse Strain L sublines, NCTC clone 929 (L929 cells) in a serum-free environment. Gibco™ Waymouth's Medium is also used for the culture of whole organs and a broad range of cell types, such as carcinoma cell lines.Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.
Dual-Site cGMP Manufacturing
Gibco™ Waymouth's Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Waymouth's Medium product made in our Grand Island facility (11220-035). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Waymouth's Medium contains no proteins, lipids, or growth factors. Some cell types may required the supplementation with Fetal Bovine Serum (FBS). Waymouth's Medium uses a sodium bicarbonate buffer system (2.24 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
Waymouth's Medium, 500mL, Liquid Form, With L-glutamine, Phenol Red, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture | |
Waymouth's Medium | |
No HEPES | |
For in-vitro diagnostic use |
2 to 8°C | |
Liquid | |
Glutamine, Phenol Red | |
Mammalian cell culture |
Safety and Handling
missing translation for 'shelfLife' : 12 Months