Learn More
Watson-Marlow 313 Rapid-Load Pumphead Extension
Offers simple, error-free tube loading
Brand:  Watson-Marlow 033.3431.000
307.46 EUR valid until 2024-11-30
Use promo code "21681" to get your promotional price.
Product Code. 10601881
  • 381.00€ / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Description
  • Pumphead accepts seven tube sizes to meet any flow requirement
  • Three nylatron rollers
  • Tube wall of 1.6mm
  • With adjustable tube clamps
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Metering and transfer pump
Adjustable tube clamps
Extension Pump Head
3
Nylatron