Learn More
LABPLAS Water Sampling Kit Writing
Sterile water sampling bag with writing area for easy labeling.
53.30€ - 90.60€
Specifications
Color Clear with White Block
View More Specs
Products 4
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
17910245
View Documents
LABPLAS
KWS-21100
100 mL
53.30€
Pack of 100
17980225
View Documents
LABPLAS
KWS-22200
500 mL
64.60€
Pack of 100
17930115
View Documents
LABPLAS
KWS-33100
300 mL
61.85€
Pack of 100
17980135
View Documents
LABPLAS
KWS-34200
1000 mL
90.60€
Pack of 100
Description
Description
  • Water Sampling Kit
  • Includes sodium thiosulfate tablet
  • Clear, with writing area for easy labeling
Specifications
Specifications
Clear with White Block