missing translation for 'onlineSavingsMsg'
Learn More

Melford Water Molecular Biology Grade DNase and RNase Free

Certified RNase and DNase free for use in molecular biology applications.

58.15€ - 87.65€

Specifications

Chemical Name or Material Water, Molecular Biology Grade, DNase and RNase Free
CAS 7732-18-5
Molecular Weight (g/mol) 18
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30017923
View Documents
Melford
248700
1 gal.
87.65€
4 L
Please to purchase this item. Need a web account? Register with us today!
30017924
View Documents
Melford
248798
1 L
58.15€
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Certified RNase and DNase free for use in molecular biology applications.

Certified RNase and DNase free for use in molecular biology applications. Ideal for making reagents, rinsing glass and plastic ware, enzymatic reactions and more. Toxic agents such as DEPC are not used in purification procedures.

  • Endotoxin Free
  • RNase Free
  • DNase Free

Tested to < 0.001 EU/ML

Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water.

Specifications

Specifications

Water, Molecular Biology Grade, DNase and RNase Free
7732-18-5
18
Videos
SDS
Documents

Documents

Product Certifications